Product Details

SNP ID
rs200396175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176130705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGCCGCCCCCCCTCCTCCTCC[G/T]CATCCTCCGCCTCCGCCGCCACCTC
Phenotype
MIM: 142980 MIM: 142985 MIM: 142982
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HOXD-AS2 PubMed Links

Gene Details

Gene
HOXD-AS2
Gene Name
HOXD cluster antisense RNA 2
There are no transcripts associated with this gene.

Gene
HOXD3
Gene Name
homeobox D3
There are no transcripts associated with this gene.

Gene
HOXD8
Gene Name
homeobox D8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199746.1 966 Silent Mutation CCG,CCT P113P NP_001186675.1
NM_001199747.1 966 Intron NP_001186676.1
NM_019558.3 966 Silent Mutation CCG,CCT P113P NP_062458.1
Gene
HOXD9
Gene Name
homeobox D9
There are no transcripts associated with this gene.

Gene
LOC100129455
Gene Name
uncharacterized LOC100129455
There are no transcripts associated with this gene.

View Full Product Details