Product Details

SNP ID
rs200425007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218383024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCATCTCCAGCCCGACCAGCCC[A/G]ACCAGCCCAGGGCCACAGCAAGCAC
Phenotype
MIM: 600266
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC11A1 PubMed Links

Gene Details

Gene
SLC11A1
Gene Name
solute carrier family 11 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000578.3 675 Silent Mutation CCA,CCG P24P NP_000569.3
XM_005246793.3 675 Intron XP_005246850.1
XM_005246794.3 675 UTR 5 XP_005246851.1
XM_006712709.3 675 Intron XP_006712772.1
XM_006712710.3 675 Intron XP_006712773.1
XM_006712711.3 675 Intron XP_006712774.1
XM_011511684.2 675 UTR 5 XP_011509986.1
XM_011511685.2 675 Intron XP_011509987.1
XM_017004765.1 675 Silent Mutation CCA,CCG P24P XP_016860254.1
XM_017004766.1 675 UTR 5 XP_016860255.1
XM_017004767.1 675 Silent Mutation CCA,CCG P24P XP_016860256.1

View Full Product Details