Product Details

SNP ID
rs201796657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166198940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTGCCTTAAGCGGTAACGTCTA[A/T]AAGCACGCTGAATGACAGTAGCAGA
Phenotype
MIM: 603415
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 5974 Missense Mutation TAT,TTT Y1889F NP_002968.1
XM_005246757.2 5974 Missense Mutation TAT,TTT Y1900F XP_005246814.1
XM_011511616.2 5974 Missense Mutation TAT,TTT Y1900F XP_011509918.1
XM_011511617.2 5974 Missense Mutation TAT,TTT Y1900F XP_011509919.1
XM_011511618.2 5974 Missense Mutation TAT,TTT Y1889F XP_011509920.1
XM_011511619.2 5974 Intron XP_011509921.1
XM_017004668.1 5974 Missense Mutation TAT,TTT Y1771F XP_016860157.1
XM_017004669.1 5974 Missense Mutation TAT,TTT Y1652F XP_016860158.1

View Full Product Details