Product Details

SNP ID
rs201760629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32633950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCAACTGGACGGGGCCCCCTG[C/T]TGACTTACACCCTCAGGACTTTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4765 PubMed Links

Gene Details

Gene
MIR4765
Gene Name
microRNA 4765
There are no transcripts associated with this gene.

Gene
TTC27
Gene Name
tetratricopeptide repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193509.1 614 Missense Mutation GCT,GTT A64V NP_001180438.1
NM_017735.4 614 Missense Mutation GCT,GTT A114V NP_060205.3
XM_005264416.1 614 Missense Mutation GCT,GTT A114V XP_005264473.1
XM_011532958.1 614 Missense Mutation GCT,GTT A114V XP_011531260.1

View Full Product Details