Product Details

SNP ID
rs202001688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113006076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATAGCAGTCCCGAGGAAGGACC[A/G]TATGTCTCCAGGTGAGTAGCCACGG
Phenotype
MIM: 605509
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL36A PubMed Links

Gene Details

Gene
IL36A
Gene Name
interleukin 36, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014440.2 524 Missense Mutation CAT,CGT H38R NP_055255.1
XM_005263639.2 524 Missense Mutation CAT,CGT H38R XP_005263696.1
XM_011510965.1 524 Missense Mutation CAT,CGT H38R XP_011509267.1
XM_017003806.1 524 Missense Mutation CAT,CGT H38R XP_016859295.1

View Full Product Details