Product Details
- SNP ID
-
rs72503549
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:75730618 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCAGAGTTCACAAGAGAGATCAAA[C/G]CTGGAGATAAAAGATTTTGAAACTA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4273
PubMed Links
Gene Details
- Gene
- MIR4273
- Gene Name
- microRNA 4273
There are no transcripts associated with this gene.
- Gene
- ZNF717
- Gene Name
- zinc finger protein 717
There are no transcripts associated with this gene.
View Full Product Details