Product Details

SNP ID
rs202033675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:142962503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCACCAGCCAGAAGTAGCGGCG[A/G]TAGAAGTGCACGAAGAGTGTGGGGT
Phenotype
MIM: 614580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAQR9 PubMed Links

Gene Details

Gene
PAQR9
Gene Name
progestin and adipoQ receptor family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198504.2 834 Silent Mutation TAC,TAT Y278Y NP_940906.1
Gene
PAQR9-AS1
Gene Name
PAQR9 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details