Product Details

SNP ID
rs200679569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51934726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCCCGCAGCCCGTGAGCTTCA[C/T]GCTGCAGGGCAAGTGGTCGCTTCTT
Phenotype
MIM: 607726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP3 PubMed Links

Gene Details

Gene
PARP3
Gene Name
poly(ADP-ribose) polymerase family member 3
There are no transcripts associated with this gene.

Gene
RRP9
Gene Name
ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004704.4 1159 Missense Mutation CAT,CGT H362R NP_004695.1

View Full Product Details