Product Details

SNP ID
rs200719486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138946369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGCCCTTGCGCTCGCCGCCGCC[C/T]TCGCGCGGCACCTTGATGAAGCACT
Phenotype
MIM: 605597
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXL2 PubMed Links

Gene Details

Gene
FOXL2
Gene Name
forkhead box L2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023067.3 772 Silent Mutation GAA,GAG E118E NP_075555.1
Gene
FOXL2NB
Gene Name
FOXL2 neighbor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040061.2 772 Intron NP_001035150.1
XM_005247443.3 772 Intron XP_005247500.1
Gene
LINC01391
Gene Name
long intergenic non-protein coding RNA 1391
There are no transcripts associated with this gene.

View Full Product Details