Product Details
- SNP ID
-
rs201166256
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:96814700 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGTCCTCCTCCGAAGCAGCTGCAC[A/C]TGCAACTGGGCAGCCTGGACCCTCG
- Phenotype
-
MIM: 600066
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EPHA6
PubMed Links
Gene Details
- Gene
- EPHA6
- Gene Name
- EPH receptor A6
View Full Product Details