Product Details

SNP ID
rs200927099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131462788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCATCATACAAATCTTCTGGCA[A/G]TTCCTCTTCATCTCCATCAGGAAAA
Phenotype
MIM: 607118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL3 PubMed Links

Gene Details

Gene
MRPL3
Gene Name
mitochondrial ribosomal protein L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007208.3 1177 Silent Mutation CTG,TTG L328L NP_009139.1

View Full Product Details