Product Details

SNP ID
rs199805758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150552370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTCTTTAGTCCGAGGATCAGAA[A/G]GACTGTACATGGTGAATGGACCACC
Phenotype
MIM: 609234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF2A PubMed Links

Gene Details

Gene
EIF2A
Gene Name
eukaryotic translation initiation factor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319043.1 168 Missense Mutation AGA,GGA R15G NP_001305972.1
NM_001319044.1 168 Missense Mutation AGA,GGA R15G NP_001305973.1
NM_001319045.1 168 Missense Mutation AGA,GGA R15G NP_001305974.1
NM_001319046.1 168 UTR 5 NP_001305975.1
NM_032025.4 168 Missense Mutation AGA,GGA R15G NP_114414.2
XM_011513224.1 168 Missense Mutation AGA,GGA R15G XP_011511526.1
Gene
SERP1
Gene Name
stress-associated endoplasmic reticulum protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014445.3 168 Intron NP_055260.1
XM_011512698.2 168 Intron XP_011511000.1

View Full Product Details