Product Details

SNP ID
rs200641668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131468124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAATTTTTATGTCCAGGTACAGA[A/G]CCATTTACATAGATTATGTTGTGCT
Phenotype
MIM: 607118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL3 PubMed Links

Gene Details

Gene
MRPL3
Gene Name
mitochondrial ribosomal protein L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007208.3 1056 Silent Mutation GGC,GGT G287G NP_009139.1

View Full Product Details