Product Details

SNP ID
rs200475552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1693684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCACTTGACTGTCCATGTGTCT[C/G]ACCTTGGTGGGTGTGCCAGAGTACA
Phenotype
MIM: 602422
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM53A PubMed Links

Gene Details

Gene
FAM53A
Gene Name
family with sequence similarity 53 member A
There are no transcripts associated with this gene.

Gene
SLBP
Gene Name
stem-loop binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306074.1 1279 Silent Mutation GTC,GTG V207V NP_001293003.1
NM_001306075.1 1279 Silent Mutation GTC,GTG V203V NP_001293004.1
NM_006527.3 1279 Silent Mutation GTC,GTG V242V NP_006518.1

View Full Product Details