Product Details

SNP ID
rs199618373
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:74750714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCTTGCTCCAATGTAGCCTTCA[C/T]CACAGCTATAAAACAAGACGAGGGC
Phenotype
MIM: 600345
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
BTC PubMed Links

Gene Details

Gene
BTC
Gene Name
betacellulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316963.1 629 Intron NP_001303892.1
NM_001729.3 629 Missense Mutation GAT,GGT D96G NP_001720.1
XM_011532211.1 629 Missense Mutation GAT,GGT D96G XP_011530513.1
XM_017008568.1 629 Intron XP_016864057.1

View Full Product Details