Product Details

SNP ID
rs199701436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139266834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTCCGTGGTTTCCCTCGCCGCT[C/G]CATCTGTGACCTCTGGCCCGGTTTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MGARP PubMed Links

Gene Details

Gene
MGARP
Gene Name
mitochondria localized glutamic acid rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032623.3 668 Missense Mutation GCA,GGA A163G NP_116012.2

View Full Product Details