Product Details

SNP ID
rs201508693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:89113752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAAAGGTGCATGGATAGAACAG[C/T]CTGTTTTCAGACAGTGGTTTGAAAA
Phenotype
MIM: 613299 MIM: 612973
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM13A PubMed Links

Gene Details

Gene
FAM13A
Gene Name
family with sequence similarity 13 member A
There are no transcripts associated with this gene.

Gene
TIGD2
Gene Name
tigger transposable element derived 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145715.2 936 Missense Mutation CCT,TCT P260S NP_663761.1

View Full Product Details