Product Details

SNP ID
rs11557156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:72223848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCAGCACTGCACCCAGCACATC[G/A]AGCTGTGGAGCAGAAAGAGGGTCAG
Phenotype
MIM: 611989
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MRPS27 PubMed Links

Gene Details

Gene
MRPS27
Gene Name
mitochondrial ribosomal protein S27
There are no transcripts associated with this gene.

View Full Product Details