Product Details

SNP ID
rs200556076
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:72223835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAAGTCAGAGCCTTCAGCACTG[C/T]ACCCAGCACATCGAGCTGTGGAGCA
Phenotype
MIM: 611989
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MRPS27 PubMed Links
Additional Information
For this assay, SNP(s) [rs3209157] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MRPS27
Gene Name
mitochondrial ribosomal protein S27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286748.1 857 Missense Mutation ACA,GCA T299A NP_001273677.1
NM_001286751.1 857 Missense Mutation ACA,GCA T229A NP_001273680.1
NM_015084.2 857 Missense Mutation ACA,GCA T285A NP_055899.2

View Full Product Details