Product Details

SNP ID
rs200852425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43291188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTTCTGCTGCTGTGGCAGGGAG[C/T]CTTGGTACTTTCTTGGCAGGGCTTG
Phenotype
MIM: 142940
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMGCS1 PubMed Links

Gene Details

Gene
HMGCS1
Gene Name
3-hydroxy-3-methylglutaryl-CoA synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098272.2 1626 Silent Mutation NP_001091742.1
NM_001324219.1 1626 Silent Mutation NP_001311148.1
NM_001324220.1 1626 Silent Mutation NP_001311149.1
NM_001324222.1 1626 Silent Mutation NP_001311151.1
NM_001324223.1 1626 Silent Mutation NP_001311152.1
NM_001324224.1 1626 Silent Mutation NP_001311153.1
NM_002130.7 1626 Silent Mutation NP_002121.4
XM_011514036.2 1626 Silent Mutation XP_011512338.1
XM_017009412.1 1626 Silent Mutation XP_016864901.1

View Full Product Details