Product Details

SNP ID
rs202125160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:99921039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGTTTCTGGAAATTTCCACTCAG[A/C]AGGATGTAGAGAAAAGGGTTAATGC
Phenotype
MIM: 606111
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MCHR2 PubMed Links

Gene Details

Gene
MCHR2
Gene Name
melanin concentrating hormone receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040179.1 1239 Silent Mutation CTG,CTT L308L NP_001035269.1
NM_032503.2 1239 Silent Mutation CTG,CTT L308L NP_115892.2

View Full Product Details