Product Details
- SNP ID
-
rs201737573
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:26240470 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGTAAAGGTTTAGGAAAGGGAGG[A/C]GCCAAGCGCCATCGCAAAGTGCTGC
- Phenotype
-
MIM: 142210
MIM: 602816
MIM: 602824
MIM: 602832
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HIST1H1D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3734533] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HIST1H1D
- Gene Name
- histone cluster 1, H1d
There are no transcripts associated with this gene.
- Gene
- HIST1H3F
- Gene Name
- histone cluster 1, H3f
There are no transcripts associated with this gene.
- Gene
- HIST1H4F
- Gene Name
- histone cluster 1, H4f
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003540.3 |
45 |
Silent Mutation |
GGA,GGC |
G15G |
NP_003531.1 |
- Gene
- HIST1H4G
- Gene Name
- histone cluster 1, H4g
There are no transcripts associated with this gene.
View Full Product Details