Product Details

SNP ID
rs201454152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43640048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAGATGCTGAAGAAGAAACCT[C/T]GGGCCACCACTGAGGTGAGCAGCAG
Phenotype
MIM: 612648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAD2L1BP PubMed Links

Gene Details

Gene
MAD2L1BP
Gene Name
MAD2L1 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003690.1 376 Missense Mutation CGG,TGG R146W NP_001003690.1
NM_014628.2 376 Missense Mutation CGG,TGG R114W NP_055443.1
Gene
RSPH9
Gene Name
radial spoke head 9 homolog
There are no transcripts associated with this gene.

View Full Product Details