Product Details

SNP ID
rs200378080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109465895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCGTTGGCTGGGGCACGTGGT[A/G]AGTGGGTGGTGGCGGACCAGGCTCC
Phenotype
MIM: 607129 MIM: 614064
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MICAL1 PubMed Links

Gene Details

Gene
MICAL1
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159291.1 2218 Intron NP_001152763.1
NM_001286613.1 2218 UTR 5 NP_001273542.1
NM_022765.3 2218 Intron NP_073602.3
Gene
ZBTB24
Gene Name
zinc finger and BTB domain containing 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164313.1 2218 Intron NP_001157785.1
NM_014797.2 2218 Missense Mutation CAC,TAC H684Y NP_055612.2

View Full Product Details