Product Details

SNP ID
rs200305624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:137493790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGATGTAGTTTCTGGCGAGCAG[C/G]AGTGTGGCGATCTTGGAGAGCTTGC
Phenotype
MIM: 609323
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OLIG3 PubMed Links

Gene Details

Gene
OLIG3
Gene Name
oligodendrocyte transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175747.2 605 Silent Mutation CTC,CTG L127L NP_786923.1

View Full Product Details