Product Details

SNP ID
rs201067708
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100101719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTACTCTCCAGAACGGCCATGA[C/T]TTCCCAATTCTTCATTCTGTCCTCC
Phenotype
MIM: 602296 MIM: 614729 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984 MIM: 602955
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AP4M1 PubMed Links

Gene Details

Gene
AP4M1
Gene Name
adaptor related protein complex 4 mu 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004722.3 222 Missense Mutation ACT,ATT T2I NP_004713.2
XM_005250689.4 222 Missense Mutation ACT,ATT T2I XP_005250746.1
XM_005250690.4 222 Missense Mutation ACT,ATT T2I XP_005250747.1
XM_006716175.3 222 Missense Mutation ACT,ATT T2I XP_006716238.1
XM_011516685.2 222 Missense Mutation ACT,ATT T2I XP_011514987.1
XM_017012790.1 222 Intron XP_016868279.1
XM_017012791.1 222 UTR 5 XP_016868280.1
Gene
COPS6
Gene Name
COP9 signalosome subunit 6
There are no transcripts associated with this gene.

Gene
MCM7
Gene Name
minichromosome maintenance complex component 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278595.1 222 UTR 5 NP_001265524.1
NM_005916.4 222 UTR 5 NP_005907.3
NM_182776.2 222 Intron NP_877577.1
XM_005250348.3 222 Intron XP_005250405.1
XM_017012217.1 222 Intron XP_016867706.1
Gene
MIR106B
Gene Name
microRNA 106b
There are no transcripts associated with this gene.

Gene
MIR25
Gene Name
microRNA 25
There are no transcripts associated with this gene.

Gene
MIR93
Gene Name
microRNA 93
There are no transcripts associated with this gene.

Gene
TAF6
Gene Name
TATA-box binding protein associated factor 6
There are no transcripts associated with this gene.

View Full Product Details