Product Details

SNP ID
rs200424612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130733446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGTGCAGCCCGGGAAGGGGCA[G/T]TGGTGGCGCTTGGCAGCAGGTGTGA
Phenotype
MIM: 609393
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLF14 PubMed Links

Gene Details

Gene
KLF14
Gene Name
Kruppel like factor 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138693.3 588 Silent Mutation NP_619638.2

View Full Product Details