Product Details

SNP ID
rs201841970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:97024287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTACCTGGCTGGTTGGTGGCGC[C/T]TGGGACGCGGTTGTAGGCGCCGCCG
Phenotype
MIM: 600028
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DLX5 PubMed Links

Gene Details

Gene
DLX5
Gene Name
distal-less homeobox 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005221.5 545 Missense Mutation AGC,GGC S113G NP_005212.1
XM_005250185.3 545 Intron XP_005250242.1
XM_017011803.1 545 Intron XP_016867292.1

View Full Product Details