Product Details
- SNP ID
-
hCV191827525
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:130733141 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTTCCACCTCGCTGGTGAGTGGC[G/T]GGTCGATGCGGGGAGTTCGACGACG
- Phenotype
-
MIM: 609393
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KLF14
PubMed Links
Gene Details
- Gene
- KLF14
- Gene Name
- Kruppel like factor 14
There are no transcripts associated with this gene.
View Full Product Details