Product Details

SNP ID
rs201137000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:94400232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCCAGGCAGAGATGGTGAAGAT[A/G]GTCCCACAGGCCCTCCTGGTCCACC
Phenotype
MIM: 120160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL1A2 PubMed Links

Gene Details

Gene
COL1A2
Gene Name
collagen type I alpha 2 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000089.3 640 Missense Mutation AGT,GGT S57G NP_000080.2

View Full Product Details