Product Details

SNP ID
rs199863923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103300206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAGAAGAGATCCCAGTTAGATC[C/T]GGAACTTGAGATTGAAAATATGGGT
Phenotype
MIM: 603131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PMPCB PubMed Links

Gene Details

Gene
PMPCB
Gene Name
peptidase, mitochondrial processing beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004279.2 390 Missense Mutation CCG,CTG P119L NP_004270.2
XM_005250717.3 390 Missense Mutation CCG,CTG P119L XP_005250774.1
XM_006716181.3 390 Missense Mutation CCG,CTG P119L XP_006716244.1

View Full Product Details