Product Details

SNP ID
rs201435015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143613599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGATGGAGACCTCATCTTCCTC[A/G]CCCACTGTGGGGAGCCACCGGGTCA
Phenotype
MIM: 616408 MIM: 137020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PYCRL PubMed Links

Gene Details

Gene
PYCRL
Gene Name
pyrroline-5-carboxylate reductase-like
There are no transcripts associated with this gene.

Gene
TSTA3
Gene Name
tissue specific transplantation antigen P35B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317783.1 862 Silent Mutation GGC,GGT G251G NP_001304712.1
NM_003313.3 862 Silent Mutation GGC,GGT G245G NP_003304.1
XM_005251051.3 862 Silent Mutation GGC,GGT G245G XP_005251108.2
XM_011517269.1 862 Silent Mutation GGC,GGT G245G XP_011515571.1

View Full Product Details