Product Details

SNP ID
rs200581623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144083198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCGCTGACCCAGCGCACTTACA[A/T]GTCGGAGAACGCCATGGGCTCCACC
Phenotype
MIM: 606491 MIM: 603048
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EXOSC4 PubMed Links

Gene Details

Gene
EXOSC4
Gene Name
exosome component 4
There are no transcripts associated with this gene.

Gene
GPAA1
Gene Name
glycosylphosphatidylinositol anchor attachment 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003801.3 259 Missense Mutation AAG,ATG K50M NP_003792.1
Gene
MIR6847
Gene Name
microRNA 6847
There are no transcripts associated with this gene.

View Full Product Details