Product Details

SNP ID
rs200392542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:21077529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGTCTTCAGATGGTTTATCTGA[C/T]GATAGACATTAGCCAGGAGGTTCTC
Phenotype
MIM: 147640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFNB1 PubMed Links

Gene Details

Gene
IFNB1
Gene Name
interferon beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002176.3 435 Missense Mutation CAT,CGT H114R NP_002167.1

View Full Product Details