Product Details

SNP ID
rs200976346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:72056840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGTTTTGTCTGGCAGGTTCCC[A/G]GGTCTAAAAGACATCCATGGAAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf57 PubMed Links

Gene Details

Gene
C9orf57
Gene Name
chromosome 9 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128618.1 107 Missense Mutation CCG,CTG P56L NP_001122090.1
XM_011518225.2 107 Missense Mutation CCG,CTG P6L XP_011516527.1

View Full Product Details