Product Details

SNP ID
rs199892252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129739859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTCTCCATGTCGTTCCGGTGGG[C/T]CCTGGGGAGACAAGAGGGGTGCGGT
Phenotype
MIM: 605172
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGES PubMed Links

Gene Details

Gene
PTGES
Gene Name
prostaglandin E synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004878.4 625 Missense Mutation ACC,GCC T71A NP_004869.1
XM_011519210.2 625 Missense Mutation ACC,GCC T99A XP_011517512.1

View Full Product Details