Product Details

SNP ID
rs199739092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34621499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGGGGCCCTGTGGAGGGTGGGG[A/G]TGCAGGGTTGGTTGGACCCTGGGAA
Phenotype
MIM: 607387
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARID3C PubMed Links

Gene Details

Gene
ARID3C
Gene Name
AT-rich interaction domain 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017363.1 4284 Missense Mutation CCC,TCC P400S NP_001017363.1
XM_017014283.1 4284 Missense Mutation CCC,TCC P550S XP_016869772.1
Gene
DCTN3
Gene Name
dynactin subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281425.1 4284 Intron NP_001268354.1
NM_001281426.1 4284 Intron NP_001268355.1
NM_001281427.1 4284 Intron NP_001268356.1
NM_007234.4 4284 Intron NP_009165.1
NM_024348.3 4284 Intron NP_077324.1
Gene
RPP25L
Gene Name
ribonuclease P/MRP subunit p25 like
There are no transcripts associated with this gene.

View Full Product Details