Product Details
- SNP ID
-
rs201905471
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:85547249 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCACTGTAATCAACTTCTTCAAGGA[A/G]TCGAGGTTCATCTTCATCCAAGACA
- Phenotype
-
MIM: 606830
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AGTPBP1
PubMed Links
Gene Details
- Gene
- AGTPBP1
- Gene Name
- ATP/GTP binding protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286715.1 |
3642 |
Missense Mutation |
CTC,TTC |
L1233F |
NP_001273644.1 |
NM_001286717.1 |
3642 |
Missense Mutation |
CTC,TTC |
L1193F |
NP_001273646.1 |
NM_015239.2 |
3642 |
Missense Mutation |
CTC,TTC |
L1141F |
NP_056054.2 |
XM_005251847.3 |
3642 |
Missense Mutation |
CTC,TTC |
L1181F |
XP_005251904.1 |
XM_005251848.2 |
3642 |
Missense Mutation |
CTC,TTC |
L1181F |
XP_005251905.1 |
XM_005251849.3 |
3642 |
Missense Mutation |
CTC,TTC |
L1063F |
XP_005251906.1 |
XM_011518418.2 |
3642 |
Intron |
|
|
XP_011516720.1 |
XM_011518420.2 |
3642 |
Missense Mutation |
CTC,TTC |
L826F |
XP_011516722.1 |
XM_011518421.2 |
3642 |
Intron |
|
|
XP_011516723.1 |
XM_017014544.1 |
3642 |
Missense Mutation |
CTC,TTC |
L1181F |
XP_016870033.1 |
XM_017014545.1 |
3642 |
Intron |
|
|
XP_016870034.1 |
XM_017014546.1 |
3642 |
Missense Mutation |
CTC,TTC |
L1023F |
XP_016870035.1 |
XM_017014547.1 |
3642 |
Intron |
|
|
XP_016870036.1 |
XM_017014548.1 |
3642 |
Missense Mutation |
CTC,TTC |
L916F |
XP_016870037.1 |
XM_017014549.1 |
3642 |
Intron |
|
|
XP_016870038.1 |
XM_017014550.1 |
3642 |
Intron |
|
|
XP_016870039.1 |
- Gene
- LOC101927603
- Gene Name
- uncharacterized LOC101927603
There are no transcripts associated with this gene.
View Full Product Details