Product Details

SNP ID
rs202209674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120534550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCATTAAACATTAGCAGCACCA[A/G]TGTTTGAAAAAGAGAGACCTGGAGT
Phenotype
MIM: 300304
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUL4B PubMed Links

Gene Details

Gene
CUL4B
Gene Name
cullin 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079872.1 2521 Silent Mutation CTG,TTG L733L NP_001073341.1
NM_003588.3 2521 Silent Mutation CTG,TTG L751L NP_003579.3
XM_005262481.1 2521 Silent Mutation CTG,TTG L753L XP_005262538.1
XM_006724784.1 2521 Silent Mutation CTG,TTG L740L XP_006724847.1
XM_006724785.2 2521 Silent Mutation CTG,TTG L738L XP_006724848.1
XM_011531399.2 2521 Silent Mutation CTG,TTG L557L XP_011529701.1
XM_011531400.2 2521 Silent Mutation CTG,TTG L555L XP_011529702.1
XM_011531401.1 2521 Silent Mutation CTG,TTG L539L XP_011529703.1

View Full Product Details