Product Details

SNP ID
rs202243976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:104040397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGAGGCCCAGGGCACGAATGCC[A/G]CCCTCAGGTACACCAAAAGCAAGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
H2BFM PubMed Links

Gene Details

Gene
H2BFM
Gene Name
H2B histone family member M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164416.1 645 Missense Mutation ACC,GCC T141A NP_001157888.1
XM_006724640.2 645 Missense Mutation ACC,GCC T141A XP_006724703.1
XM_011530922.2 645 Missense Mutation ACC,GCC T141A XP_011529224.1

View Full Product Details