Product Details
- SNP ID
-
rs202062315
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:20015302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGACGCATCTGTTCAGCTACCTCC[C/T]GGGCCTTTGTTTCTGCTGCTTCTTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAP7D2
PubMed Links
Gene Details
- Gene
- MAP7D2
- Gene Name
- MAP7 domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001168465.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q557R |
NP_001161937.1 |
NM_001168466.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q471R |
NP_001161938.1 |
NM_001168467.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q464R |
NP_001161939.1 |
NM_152780.3 |
1559 |
Missense Mutation |
CAG,CGG |
Q516R |
NP_689993.2 |
XM_005274478.2 |
1559 |
Missense Mutation |
CAG,CGG |
Q541R |
XP_005274535.1 |
XM_005274479.2 |
1559 |
Missense Mutation |
CAG,CGG |
Q530R |
XP_005274536.1 |
XM_005274480.3 |
1559 |
Missense Mutation |
CAG,CGG |
Q508R |
XP_005274537.1 |
XM_006724481.2 |
1559 |
Missense Mutation |
CAG,CGG |
Q590R |
XP_006724544.1 |
XM_006724482.2 |
1559 |
Missense Mutation |
CAG,CGG |
Q549R |
XP_006724545.1 |
XM_011545483.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q579R |
XP_011543785.1 |
XM_011545484.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q546R |
XP_011543786.1 |
XM_011545485.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q535R |
XP_011543787.1 |
XM_011545486.2 |
1559 |
Intron |
|
|
XP_011543788.1 |
XM_017029393.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q538R |
XP_016884882.1 |
XM_017029394.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q505R |
XP_016884883.1 |
XM_017029395.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q497R |
XP_016884884.1 |
XM_017029396.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q494R |
XP_016884885.1 |
XM_017029397.1 |
1559 |
Missense Mutation |
CAG,CGG |
Q486R |
XP_016884886.1 |
XM_017029398.1 |
1559 |
Intron |
|
|
XP_016884887.1 |
- Gene
- MIR23C
- Gene Name
- microRNA 23c
There are no transcripts associated with this gene.
View Full Product Details