Product Details
- SNP ID
-
rs201220623
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:44873631 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGATGAGGAAAAGAAAATGGCGG[C/G]GGGAAAAGCGAGCGGCGAGAGCGAG
- Phenotype
-
MIM: 300128
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KDM6A
PubMed Links
Gene Details
- Gene
- KDM6A
- Gene Name
- lysine demethylase 6A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001291415.1 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
NP_001278344.1 |
NM_001291416.1 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
NP_001278345.1 |
NM_001291417.1 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
NP_001278346.1 |
NM_001291418.1 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
NP_001278347.1 |
NM_001291421.1 |
457 |
UTR 5 |
|
|
NP_001278350.1 |
NM_021140.3 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
NP_066963.2 |
XM_005272656.4 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_005272713.1 |
XM_005272659.4 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_005272716.1 |
XM_011543957.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542259.1 |
XM_011543958.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542260.1 |
XM_011543959.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542261.1 |
XM_011543960.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542262.1 |
XM_011543961.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542263.1 |
XM_011543962.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542264.1 |
XM_011543963.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542265.1 |
XM_011543964.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542266.1 |
XM_011543965.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542267.1 |
XM_011543966.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542268.1 |
XM_011543967.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542269.1 |
XM_011543968.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542270.1 |
XM_011543969.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542271.1 |
XM_011543970.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542272.1 |
XM_011543971.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542273.1 |
XM_011543972.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542274.1 |
XM_011543973.2 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542275.1 |
XM_011543974.1 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_011542276.1 |
XM_011543975.2 |
457 |
Intron |
|
|
XP_011542277.1 |
XM_017029782.1 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_016885271.1 |
XM_017029783.1 |
457 |
Missense Mutation |
GCG,GGG |
A27G |
XP_016885272.1 |
XM_017029784.1 |
457 |
UTR 5 |
|
|
XP_016885273.1 |
XM_017029785.1 |
457 |
UTR 5 |
|
|
XP_016885274.1 |
View Full Product Details