Product Details

SNP ID
rs200713134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49206588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAATCCTCACAACTGCCCTGGC[A/G]CTGCAGACACTGGATGGTGAAGGAG
Phenotype
MIM: 300110 MIM: 313475
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNA1F PubMed Links

Gene Details

Gene
CACNA1F
Gene Name
calcium voltage-gated channel subunit alpha1 F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256789.2 2663 Missense Mutation CGC,TGC R1799C NP_001243718.1
NM_001256790.2 2663 Missense Mutation CGC,TGC R1745C NP_001243719.1
NM_005183.3 2663 Missense Mutation CGC,TGC R1810C NP_005174.2
XM_011543983.2 2663 Missense Mutation CGC,TGC R1738C XP_011542285.1
XM_017029836.1 2663 Missense Mutation CGC,TGC R860C XP_016885325.1
Gene
SYP
Gene Name
synaptophysin
There are no transcripts associated with this gene.

Gene
SYP-AS1
Gene Name
SYP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details