Product Details

SNP ID
rs35890136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32635960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTCTATGAAGGGAATTTATAAT[G/T]TATATTCTACAGAGGTTTTCATCCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR4765 PubMed Links

Gene Details

Gene
MIR4765
Gene Name
microRNA 4765
There are no transcripts associated with this gene.

Gene
TTC27
Gene Name
tetratricopeptide repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193509.1 Intron NP_001180438.1
NM_017735.4 Intron NP_060205.3
XM_005264416.1 Intron XP_005264473.1
XM_011532958.1 Intron XP_011531260.1

View Full Product Details