Product Details
- SNP ID
-
rs11845687
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:74242456 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTTTAAAACGGCCTATCCTCACC[C/G]CTTAGGCCCTAATAGGTTTTGGCTT
- Phenotype
-
MIM: 142993
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
VSX2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117447114,rs77732635] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- VSX2
- Gene Name
- visual system homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182894.2 |
|
Intron |
|
|
NP_878314.1 |
View Full Product Details