Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001031739.2 | Intron | NP_001026909.1 | |||
NM_001168530.1 | Intron | NP_001162002.1 | |||
NM_001168531.1 | Intron | NP_001162003.1 | |||
NM_024087.2 | Intron | NP_076992.1 | |||
XM_005274446.1 | Intron | XP_005274503.1 | |||
XM_011545458.1 | Intron | XP_011543760.1 | |||
XM_011545460.1 | Intron | XP_011543762.1 | |||
XM_017029283.1 | Intron | XP_016884772.1 | |||
XM_017029284.1 | Intron | XP_016884773.1 | |||
XM_017029285.1 | Intron | XP_016884774.1 | |||
XM_017029286.1 | Intron | XP_016884775.1 |