Product Details

SNP ID
rs2523733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30163738 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCATGAGCTGCCTGCCTGTACCCT[A/C]TGTGCGGGGCCGCTGGAGGATGCGG
Phenotype
MIM: 605701
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRIM10 PubMed Links

Gene Details

Gene
TRIM10
Gene Name
tripartite motif containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006778.3 533 Intron NP_006769.2
NM_052828.2 533 Intron NP_439893.2
XM_011514221.1 533 Intron XP_011512523.1
XM_011514222.2 533 Intron XP_011512524.1
XM_011514223.2 533 Intron XP_011512525.1
XM_011514225.1 533 Intron XP_011512527.1
Gene
TRIM15
Gene Name
tripartite motif containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033229.2 533 Silent Mutation CTA,CTC L18L NP_150232.2
XM_011514987.1 533 Intron XP_011513289.1
XM_011514988.2 533 UTR 5 XP_011513290.1

View Full Product Details