Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001270367.1 | 372 | Intron | NP_001257296.1 | ||
NM_001270368.1 | 372 | Intron | NP_001257297.1 | ||
NM_001270369.1 | 372 | Intron | NP_001257298.1 | ||
NM_001270370.1 | 372 | Intron | NP_001257299.1 | ||
NM_001270371.1 | 372 | Intron | NP_001257300.1 | ||
NM_001270372.1 | 372 | Intron | NP_001257301.1 | ||
NM_001270373.1 | 372 | Intron | NP_001257302.1 | ||
NM_001270374.1 | 372 | UTR 5 | NP_001257303.1 | ||
NM_025125.3 | 372 | Intron | NP_079401.2 | ||
XM_006717978.2 | 372 | UTR 5 | XP_006718041.1 | ||
XM_011540192.2 | 372 | Intron | XP_011538494.1 | ||
XM_011540193.2 | 372 | UTR 5 | XP_011538495.1 | ||
XM_017016679.1 | 372 | Intron | XP_016872168.1 | ||
XM_017016680.1 | 372 | UTR 5 | XP_016872169.1 |