Product Details

SNP ID
rs175147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:16828472 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAACAGAAATCTGAGATTGTATG[G/T]GTTATGTTTGTTATTGTTACTTGCC
Phenotype
MIM: 611674
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HSFY1P1 PubMed Links

Gene Details

Gene
HSFY1P1
Gene Name
heat shock transcription factor, Y-linked 1 pseudogene 1
There are no transcripts associated with this gene.

Gene
XKR3
Gene Name
XK related 3
There are no transcripts associated with this gene.

View Full Product Details