Product Details

SNP ID
rs12634139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128466689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCATCACCGGTATAATTCCAGA[C/T]TAGAGCCTGCCAATACAAGGGATTT
Phenotype
MIM: 611337
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB8 PubMed Links

Gene Details

Gene
DNAJB8
Gene Name
DnaJ heat shock protein family (Hsp40) member B8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153330.4 280 UTR 5 NP_699161.1
XM_006713519.3 280 UTR 5 XP_006713582.1
Gene
DNAJB8-AS1
Gene Name
DNAJB8 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details